ADVH0430A0C9 AChemBlock

2-(Piperazin-2-yl)acetonitrile dihydrochloride

  • MDL number MFCD31620760
  • Cas Number : 142054-62-4
  • Molecular Weight : 198.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Room Temp
Shipping Temperature Room Temp
Country of Origin China
Product Link https://www.achemblock.com/2-piperazin-2-yl-acetonitrile-dihydrochloride.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=K13387
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart