ADVH0430A0D1 AChemBlock

5-Azaspiro[3.4]octane hemioxalate

  • MDL number MFCD27988102
  • Cas Number : 1403766-69-7
  • Molecular Weight : 312.41

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form white solid
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/5-azaspiro-3-4-octane-oxalate.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-5104
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart