ADVH0430A0E4 AChemBlock

3-[(2,3-Dichlorophenyl)amino]propanoic acid

  • MDL number MFCD11186363
  • Cas Number : 36053-84-6
  • Molecular Weight : 234.08

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/3-2-3-dichlorophenyl-amino-propanoic-acid.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=K12434
Availability details for
Quantity
  • Availability
  • Add to Cart