ADVH0430A0E8 AChemBlock

tert-Butyl (2S,4S)-4-amino-2-methylpiperidine-1-carboxylate

  • MDL number MFCD22397554
  • Cas Number : 1628833-70-4
  • Molecular Weight : 214.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/tert-butyl-2s-4s-4-amino-2-methylpiperidine-1-carboxylate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=K12438
Availability details for
Quantity
  • Availability
  • Add to Cart