ADVH0430A114 AChemBlock

Potassium 3-(tert-butyl)-1,2,4-oxadiazole-5-carboxylate

  • MDL number MFCD31620759
  • Cas Number : 2173992-51-1
  • Molecular Weight : 208.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Room Temp
Shipping Temperature Room Temp
Country of Origin China
Product Link https://www.achemblock.com/potassium-3-tert-butyl-1-2-4-oxadiazole-5-carboxylate.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=K13384
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart