ADVH0430A130 AChemBlock

2,4-Difluorobenzeneethanol

  • MDL number MFCD00142823
  • Cas Number : 81228-02-6
  • Molecular Weight : 158.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/2-4-difluorobenzeneethanol.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=O27872
Availability details for
Quantity
  • Availability
  • Add to Cart