ADVH0430A1A1 AChemBlock

2-(3-Chlorophenoxy)-2-methyl-propanoic acid

  • MDL number MFCD01695660
  • Cas Number : 17413-73-9
  • Molecular Weight : 214.65

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/2-3-chlorophenoxy-2-methyl-propanoic-acid.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=O27985
Availability details for
Quantity
  • Availability
  • Add to Cart