ADVH0430A1A9 AChemBlock

4-(Bromomethyl)-3-fluorobenzoic acid methyl ester

  • MDL number MFCD00273345
  • Cas Number : 128577-47-9
  • Molecular Weight : 247.07

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/4-bromomethyl-3-fluorobenzoic-acid-methyl-ester.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=O27993
Availability details for
Quantity
  • Availability
  • Add to Cart