ADVH0430A1B4 AChemBlock

1-[3-(Trifluoromethyl)phenyl]cyclopropanecarboxylic acid

  • MDL number MFCD07374444
  • Cas Number : 104173-41-3
  • Molecular Weight : 230.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/1-3-trifluoromethyl-phenyl-cyclopropanecarboxylic-acid.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=O28004
Availability details for
Quantity
  • Availability
  • Add to Cart