ADVH0430A1B6 AChemBlock

1-(4-Methoxyphenyl)cyclopentanecarboxylic acid

  • MDL number MFCD00001374
  • Cas Number : 43050-28-8
  • Molecular Weight : 220.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/1-4-methoxyphenyl-cyclopentanecarboxylic-acid.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=O28006
Availability details for
Quantity
  • Availability
  • Add to Cart