ADVH0430A1C3 AChemBlock

2-(3,5-Dichlorophenoxy)acetic acid

  • MDL number MFCD04037337
  • Cas Number : 587-64-4
  • Molecular Weight : 221.04

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/2-3-5-dichlorophenoxy-acetic-acid.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=O28019
Availability details for
Quantity
  • Availability
  • Add to Cart