ADVH0430A1E4 AChemBlock

2-(4-Cyanophenoxy)acetic acid

  • MDL number MFCD01250758
  • Cas Number : 1878-82-6
  • Molecular Weight : 177.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/2-4-cyanophenoxy-acetic-acid.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=O28052
Availability details for
Quantity
  • Availability
  • Add to Cart