ADVH0430A1EA AChemBlock

2-[4-(Trifluoromethyl)phenoxy]propanoic acid

  • MDL number MFCD02684122
  • Cas Number : 69484-34-0
  • Molecular Weight : 234.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/2-4-trifluoromethyl-phenoxy-propanoic-acid.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=O28058
Availability details for
Quantity
  • Availability
  • Add to Cart