ADVH0430A1F4 AChemBlock

3-Pyridineethanol

  • MDL number MFCD00831047
  • Cas Number : 6293-56-7
  • Molecular Weight : 123.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/3-pyridineethanol.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=O28068
Availability details for
Quantity
  • Availability
  • Add to Cart