ADVH0430A2CD AChemBlock

3-Benzylmorpholine hydrochloride

  • MDL number MFCD04117744
  • Cas Number : 1172897-29-8
  • Molecular Weight : 213.71

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/3-benzylmorpholine-hydrochloride.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=O28285
Availability details for
Quantity
  • Availability
  • Add to Cart