ADVH0430A2E2 AChemBlock

3,3-Dimethyl-morpholine hydrochloride

  • MDL number MFCD11052542
  • Cas Number : 59229-64-0
  • Molecular Weight : 151.64

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/3-3-dimethyl-morpholine-hydrochloride.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=O28306
Availability details for
Quantity
  • Availability
  • Add to Cart