ADVH0430A2FE AChemBlock

2,2-dimethyl-3-phenylpropanoic acid

  • MDL number MFCD01722147
  • Cas Number : 5669-14-7
  • Molecular Weight : 178.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/a-a-dimethyl-benzenepropanoic-acid.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=O28334
Availability details for
Quantity
  • Availability
  • Add to Cart