ADVH0430A35F AChemBlock

4-(4-Bromophenyl)oxane-4-carboxylic acid

  • MDL number MFCD10693768
  • Cas Number : 1152567-60-6
  • Molecular Weight : 285.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/4-4-bromophenyl-oxane-4-carboxylic-acid.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=O28431
Availability details for
Quantity
  • Availability
  • Add to Cart