ADVH0430A4A0 AChemBlock

4-(1H-Pyrazol-1-yl)-piperidine dihydrochloride

  • MDL number MFCD11858004
  • Cas Number : 1181458-83-2
  • Molecular Weight : 224.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/4-1h-pyrazol-1-yl-piperidine-dihydrochloride.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=O28752
Availability details for
Quantity
  • Availability
  • Add to Cart