ADVH0430A4C8 AChemBlock

2-(Thiophen-2-yl)glycine

  • MDL number MFCD00005449
  • Cas Number : 21124-40-3
  • Molecular Weight : 157.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/2-thiophen-2-yl-glycine.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=O28791
Availability details for
Quantity
  • Availability
  • Add to Cart