ADVH0430A520 AChemBlock

1-Aminoindan-1-carboxylic acid

  • MDL number MFCD00800595
  • Cas Number : 3927-71-7
  • Molecular Weight : 177.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/1-aminoindan-1-carboxylic-acid.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=O28879
Availability details for
Quantity
  • Availability
  • Add to Cart