ADVH0430A545 AChemBlock

(1-Aminocyclohexyl)acetic acid

  • MDL number MFCD02258116
  • Cas Number : 37631-92-8
  • Molecular Weight : 157.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/1-aminocyclohexyl-acetic-acid.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=O28916
Availability details for
Quantity
  • Availability
  • Add to Cart