ADVH0430A55D AChemBlock

tert-Butyl 3-(methylamino)propionate

  • MDL number MFCD08275841
  • Cas Number : 143707-72-6
  • Molecular Weight : 159.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/tert-butyl-3-methylamino-propionate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=O28940
Availability details for
Quantity
  • Availability
  • Add to Cart