ADVH0430A804 AChemBlock

3-chloro-6-nitroisoquinoline

  • MDL number MFCD11044717
  • Cas Number : 760179-81-5
  • Molecular Weight : 208.61

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/3-chloro-6-nitroisoquinoline.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=O29619
Availability details for
Quantity
  • Availability
  • Add to Cart