ADVH0430A83B AChemBlock

2-chloro-5-iodopyridine-3-carbonitrile

  • MDL number MFCD09909360
  • Cas Number : 766515-34-8
  • Molecular Weight : 264.45

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Room Temp
Shipping Temperature Room Temp
Country of Origin China
Product Link https://www.achemblock.com/2-chloro-5-iodopyridine-3-carbonitrile.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=O29674
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart