ADVH0430A897 AChemBlock

1-(5-bromopyridin-2-yl)cyclopropanamine

  • MDL number MFCD11037259
  • Cas Number : 944718-22-3
  • Molecular Weight : 213.08

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/1-5-bromopyridin-2-yl-cyclopropanamine.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=O29766
Availability details for
Quantity
  • Availability
  • Add to Cart