ADVH0430A89E AChemBlock

4,7-dibromo-1H-benzo[d][1,2,3]triazole

  • MDL number MFCD11557217
  • Cas Number : 851106-90-6
  • Molecular Weight : 276.92

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/4-7-dibromo-1h-benzo-d-1-2-3-triazole.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=O29773
Availability details for
Quantity
  • Availability
  • Add to Cart