ADVH0430A8D4 AChemBlock

5-bromo-3-methoxyisoquinoline

  • MDL number MFCD19982316
  • Cas Number : 1330750-62-3
  • Molecular Weight : 238.09

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/5-bromo-3-methoxyisoquinoline.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=O29827
Availability details for
Quantity
  • Availability
  • Add to Cart