ADVH0430A90B AChemBlock

3-chloroisoquinolin-5-amine

  • MDL number MFCD09999240
  • Cas Number : 58142-49-7
  • Molecular Weight : 178.62

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/3-chloroisoquinolin-5-amine.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=O29882
Availability details for
Quantity
  • Availability
  • Add to Cart