ADVH0430A936 AChemBlock

2-(2-(prop-2-ynyloxy)ethoxy)ethanol

  • MDL number MFCD00053353
  • Cas Number : 7218-43-1
  • Molecular Weight : 144.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/2-2-prop-2-ynyloxy-ethoxy-ethanol.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=O29925
Availability details for
Quantity
  • Availability
  • Add to Cart