ADVH0430A98B AChemBlock

1H-pyrazolo[3,4-d]pyrimidin-6-amine

  • MDL number MFCD11559009
  • Cas Number : 287177-82-6
  • Molecular Weight : 135.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/1h-pyrazolo-3-4-d-pyrimidin-6-amine.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=O30010
Availability details for
Quantity
  • Availability
  • Add to Cart