ADVH0430A995 AChemBlock

3-chloro-5-nitroisoquinoline

  • MDL number MFCD09999239
  • Cas Number : 10296-47-6
  • Molecular Weight : 208.61

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/3-chloro-5-nitroisoquinoline.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=O30020
Availability details for
Quantity
  • Availability
  • Add to Cart