ADVH0430A997 AChemBlock

benzo[d]thiazol-4-ol

  • MDL number MFCD11046398
  • Cas Number : 7405-23-4
  • Molecular Weight : 151.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/benzo-d-thiazol-4-ol.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=O30022
Availability details for
Quantity
  • Availability
  • Add to Cart