ADVH0430A9CF AChemBlock

methyl 2-(2-chlorophenyl)-2-oxoacetate

  • MDL number MFCD18206259
  • Cas Number : 34966-49-9
  • Molecular Weight : 198.61

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/methyl-2-2-chlorophenyl-2-oxoacetate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=O30078
Availability details for
Quantity
  • Availability
  • Add to Cart