ADVH0430A9D9 AChemBlock

3-fluoro-4-isopropoxybenzenamine

  • MDL number MFCD11193311
  • Cas Number : 97566-69-3
  • Molecular Weight : 169.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/3-fluoro-4-isopropoxybenzenamine.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=O30088
Availability details for
Quantity
  • Availability
  • Add to Cart