ADVH0430AA40 AChemBlock

tert-butyl 4-(3-chlorophenyl)-4-cyanopiperidine-1-carboxylate

  • MDL number MFCD07779081
  • Cas Number : 553631-33-7
  • Molecular Weight : 320.82

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/tert-butyl-4-3-chlorophenyl-4-cyanopiperidine-1-carboxylate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=O30191
Availability details for
Quantity
  • Availability
  • Add to Cart