ADVH0430AA41 AChemBlock

6-chloro-1H-pyrazolo[3,4-d]pyrimidin-4(7H)-one

  • MDL number MFCD18793418
  • Cas Number : 42754-97-2
  • Molecular Weight : 170.56

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/6-chloro-1h-pyrazolo-3-4-d-pyrimidin-4-7h-one.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=O30192
Availability details for
Quantity
  • Availability
  • Add to Cart