ADVH0430AA53 AChemBlock

1-(5-fluoropyrimidin-2-yl)ethanone

  • MDL number MFCD18205684
  • Cas Number : 905587-44-2
  • Molecular Weight : 140.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/1-5-fluoropyrimidin-2-yl-ethanone.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=O30210
Availability details for
Quantity
  • Availability
  • Add to Cart