ADVH0430AA8C AChemBlock

1,4-dimethyl-1H-indazol-5-yl-5-boronic acid

  • MDL number MFCD12028655
  • Cas Number : 1310405-36-7
  • Molecular Weight : 190.01

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/1-4-dimethyl-1h-indazol-5-yl-5-boronic-acid.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=O30267
Availability details for
Quantity
  • Availability
  • Add to Cart