ADVH0430AAC6 AChemBlock

(isoquinolin-7-yl)methanol

  • MDL number MFCD11226917
  • Cas Number : 158654-76-3
  • Molecular Weight : 159.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/isoquinolin-7-yl-methanol.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=O30325
Availability details for
Quantity
  • Availability
  • Add to Cart