ADVH0430AAED AChemBlock

2-bromo-3-(bromomethyl)pyridine

  • MDL number MFCD08276299
  • Cas Number : 94446-97-6
  • Molecular Weight : 250.92

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/2-bromo-3-bromomethyl-pyridine.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=O30364
Availability details for
Quantity
  • Availability
  • Add to Cart