ADVH0430AB5F AChemBlock

quinoline-4-carbohydrazide

  • MDL number MFCD00784841
  • Cas Number : 29620-62-0
  • Molecular Weight : 187.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/quinoline-4-carbohydrazide.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=O30478
Availability details for
Quantity
  • Availability
  • Add to Cart