ADVH0430AB7F AChemBlock

methyl 5-bromo-2-methylthiazole-4-carboxylate

  • MDL number MFCD09265508
  • Cas Number : 899897-21-3
  • Molecular Weight : 236.09

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/methyl-5-bromo-2-methylthiazole-4-carboxylate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=O30510
Availability details for
Quantity
  • Availability
  • Add to Cart