ADVH0430AB8E AChemBlock

4-chloro-7-morpholinoquinazoline

  • MDL number MFCD24850028
  • Cas Number : 1334602-74-2
  • Molecular Weight : 249.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/4-chloro-7-morpholinoquinazoline.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=O30525
Availability details for
Quantity
  • Availability
  • Add to Cart