ADVH0430AB92 AChemBlock

1-(3-fluoropyridin-2-yl)ethanone

  • MDL number MFCD06659495
  • Cas Number : 87674-20-2
  • Molecular Weight : 139.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/1-3-fluoropyridin-2-yl-ethanone.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=O30529
Availability details for
Quantity
  • Availability
  • Add to Cart