ADVH0430AB9C AChemBlock

quinolin-7-amine

  • MDL number MFCD01646243
  • Cas Number : 580-19-8
  • Molecular Weight : 144.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/quinolin-7-amine.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=O30539
Availability details for
Quantity
  • Availability
  • Add to Cart