ADVH0430ABC5 AChemBlock

4-(pyridin-3-yl)piperidin-4-ol

  • MDL number MFCD04117758
  • Cas Number : 50461-59-1
  • Molecular Weight : 178.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/4-pyridin-3-yl-piperidin-4-ol.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=O30580
Availability details for
Quantity
  • Availability
  • Add to Cart