ADVH0430ABF3 AChemBlock

methyl 2-(methylthio)pyrimidine-4-carboxylate

  • MDL number MFCD11226174
  • Cas Number : 64224-67-5
  • Molecular Weight : 184.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/methyl-2-methylthio-pyrimidine-4-carboxylate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=O30627
Availability details for
Quantity
  • Availability
  • Add to Cart