ADVH0430AC15 AChemBlock

ethyl 6-bromo-1H-indazole-3-carboxylate

  • MDL number MFCD05664002
  • Cas Number : 885272-94-6
  • Molecular Weight : 269.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/ethyl-6-bromo-1h-indazole-3-carboxylate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=O30661
Availability details for
Quantity
  • Availability
  • Add to Cart