ADVH0430AC5C AChemBlock

4-(pyrimidin-2-yl)benzaldehyde

  • MDL number MFCD08056282
  • Cas Number : 77232-38-3
  • Molecular Weight : 184.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/4-pyrimidin-2-yl-benzaldehyde.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=O30732
Availability details for
Quantity
  • Availability
  • Add to Cart